KEY771305953 Key Organics / BIONET

methyl (2S)-2-amino-2-(2,4-difluorophenyl)acetate hydrochloride

  • MDL number MFCD24412534
  • Cas Number : 2061996-82-3
  • Molecular Weight : 237.63

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/665465

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/GS-7231_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart