KEY771305960 Key Organics / BIONET

methyl 4-[(1R)-1-amino-2-hydroxyethyl]benzoate hydrochloride

  • MDL number MFCD12910852
  • Cas Number : 1213380-39-2
  • Molecular Weight : 231.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/665472

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/GS-7242_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart