KEY771305961 Key Organics / BIONET

(2S)-2-amino-2-(3-chloro-2-fluorophenyl)ethan-1-ol hydrochloride

  • MDL number MFCD12758365
  • Cas Number : 1391506-22-1
  • Molecular Weight : 226.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (2S)-2-amino-2-(3-chloro-2-fluorophenyl)ethanol hydrochloride
Inchi Code 1S/C8H9ClFNO.ClH/c9-6-3-1-2-5(8(6)10)7(11)4-12;/h1-3,7,12H,4,11H2;1H/t7-;/m1./s1
Inchi Key SEZRFYYOQSQCSP-OGFXRTJISA-N

Description

Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/665473

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/GS-7244_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart