KEY771305971 Key Organics / BIONET

(2S)-2-amino-2-(4-fluoro-3-methoxyphenyl)acetic acid hydrochloride

  • MDL number MFCD28977503
  • Cas Number : 1810074-58-8
  • Molecular Weight : 235.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (S)-2-amino-2-(4-fluoro-3-methoxyphenyl)acetic acid hydrochloride
Inchi Code 1S/C9H10FNO3.ClH/c1-14-7-4-5(2-3-6(7)10)8(11)9(12)13;/h2-4,8H,11H2,1H3,(H,12,13);1H/t8-;/m0./s1
Inchi Key BMJIRSBYZJOWGS-QRPNPIFTSA-N

Description

Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/665483

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/GS-7259_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart