KEY771305974 Key Organics / BIONET

(2S)-2-amino-3-(2-hydroxyphenyl)propanoic acid hydrochloride

  • MDL number MFCD28986155
  • Cas Number : 1810074-88-4
  • Molecular Weight : 217.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/665486

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/GS-7264_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart