KEY771305978 Key Organics / BIONET

(1S)-5-chloro-2,3-dihydro-1H-inden-1-amine hydrochloride

  • MDL number MFCD22375542
  • Cas Number : 1376687-76-1
  • Molecular Weight : 204.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (1S)-5-chloro-2,3-dihydro-1H-inden-1-amine hydrochloride
Inchi Code 1S/C9H10ClN.ClH/c10-7-2-3-8-6(5-7)1-4-9(8)11;/h2-3,5,9H,1,4,11H2;1H/t9-;/m0./s1
Inchi Key JRYBHXVWIQBARN-FVGYRXGTSA-N

Description

Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/665490

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/GS-7270_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart