KEY771305980 Key Organics / BIONET

(2S)-2-amino-2-[4-chloro-3-(trifluoromethyl)phenyl]ethan-1-ol hydrochloride

  • MDL number MFCD12758208
  • Cas Number : 1810074-79-3
  • Molecular Weight : 276.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/665492

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/GS-7274_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart