KEY771305981 Key Organics / BIONET

(2S)-pentan-2-amine hydrochloride

  • MDL number MFCD28166282
  • Cas Number : 216237-52-4
  • Molecular Weight : 123.63

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (S)-pentan-2-amine hydrochloride
Inchi Code 1S/C5H13N.ClH/c1-3-4-5(2)6;/h5H,3-4,6H2,1-2H3;1H/t5-;/m0./s1
Inchi Key AMIBHXGATODRDN-JEDNCBNOSA-N

Description

Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/665493

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/GS-7278_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart