KEY771305986 Key Organics / BIONET

(2S)-2-amino-2-(2-chlorophenyl)ethan-1-ol hydrochloride

  • MDL number MFCD12758099
  • Cas Number : 213990-63-7
  • Molecular Weight : 208.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (2S)-2-amino-2-(2-chlorophenyl)ethanol hydrochloride
Inchi Code 1S/C8H10ClNO.ClH/c9-7-4-2-1-3-6(7)8(10)5-11;/h1-4,8,11H,5,10H2;1H/t8-;/m1./s1
Inchi Key FCJCVLVGGFRUIH-DDWIOCJRSA-N

Description

Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/665498

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/GS-7286_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart