KEY771305987 Key Organics / BIONET

(1S)-1-(3-fluorophenyl)ethan-1-amine hydrochloride

  • MDL number MFCD11035900
  • Cas Number : 321429-48-5
  • Molecular Weight : 175.63

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (1S)-1-(3-fluorophenyl)ethanamine hydrochloride
Inchi Code 1S/C8H10FN.ClH/c1-6(10)7-3-2-4-8(9)5-7;/h2-6H,10H2,1H3;1H/t6-;/m0./s1
Inchi Key RKALWMOKWLLQMU-RGMNGODLSA-N

Description

Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/665499

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/GS-7288_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart