KEY771306005 Key Organics / BIONET

(1R)-1-(3,4-dimethoxyphenyl)ethan-1-amine hydrochloride

  • MDL number MFCD24410973
  • Cas Number : 390815-41-5
  • Molecular Weight : 217.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (R)-1-(3,4-dimethoxyphenyl)ethan-1-amine hydrochloride
Inchi Code 1S/C10H15NO2.ClH/c1-7(11)8-4-5-9(12-2)10(6-8)13-3;/h4-7H,11H2,1-3H3;1H/t7-;/m1./s1
Inchi Key PQHFDOBOSKIMTO-OGFXRTJISA-N

Description

Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/665517

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/GS-7319_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart