KEY771306013 Key Organics / BIONET

(4R)-8-bromo-3,4-dihydro-2H-1-benzopyran-4-amine hydrochloride

  • MDL number MFCD24436022
  • Cas Number : 1810074-69-1
  • Molecular Weight : 264.55

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/665525

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/GS-7330_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart