KEY771306015 Key Organics / BIONET

4-[(1R)-1-amino-2-hydroxyethyl]phenol hydrochloride

  • MDL number MFCD18375672
  • Cas Number : 420110-45-8
  • Molecular Weight : 189.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-[(1R)-1-amino-2-hydroxyethyl]phenol hydrochloride
Inchi Code 1S/C8H11NO2.ClH/c9-8(5-10)6-1-3-7(11)4-2-6;/h1-4,8,10-11H,5,9H2;1H/t8-;/m0./s1
Inchi Key YUEKJBCPERBCDN-QRPNPIFTSA-N

Description

Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/665527

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/GS-7334_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart