KEY771306016 Key Organics / BIONET

(2R)-2-amino-2-(3-chlorophenyl)acetic acid hydrochloride

  • MDL number MFCD12910688
  • Cas Number : 37085-27-1
  • Molecular Weight : 222.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (2R)-amino(3-chlorophenyl)ethanoic acid hydrochloride
Inchi Code 1S/C8H8ClNO2.ClH/c9-6-3-1-2-5(4-6)7(10)8(11)12;/h1-4,7H,10H2,(H,11,12);1H/t7-;/m1./s1
Inchi Key YPIVOEYNISLYJX-OGFXRTJISA-N

Description

Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/665528

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/GS-7336_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart