KEY771306017 Key Organics / BIONET

(1R)-1-(2-nitrophenyl)ethan-1-amine hydrochloride

  • MDL number MFCD24411694
  • Cas Number : 1431699-56-7
  • Molecular Weight : 202.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (R)-1-(2-nitrophenyl)ethan-1-amine hydrochloride
Inchi Code 1S/C8H10N2O2.ClH/c1-6(9)7-4-2-3-5-8(7)10(11)12;/h2-6H,9H2,1H3;1H/t6-;/m1./s1
Inchi Key HFXVLGOYQIKRTO-FYZOBXCZSA-N

Description

Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/665529

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/GS-7338_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart