KEY771306023 Key Organics / BIONET

(1R)-1-(4-chloro-3-methylphenyl)ethan-1-amine hydrochloride

  • MDL number MFCD16294241
  • Cas Number : 1255306-36-5
  • Molecular Weight : 206.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/665535

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/GS-7349_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart