KEY771306024 Key Organics / BIONET

(2S)-2-amino-2-(4-methylphenyl)ethan-1-ol hydrochloride

  • MDL number MFCD12758127
  • Cas Number : 327183-90-4
  • Molecular Weight : 187.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (2S)-2-amino-2-(4-methylphenyl)ethanol hydrochloride
Inchi Code 1S/C9H13NO.ClH/c1-7-2-4-8(5-3-7)9(10)6-11;/h2-5,9,11H,6,10H2,1H3;1H/t9-;/m1./s1
Inchi Key SXKKIUVSYPXHBT-SBSPUUFOSA-N

Description

Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/665536

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/GS-7350_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart