KEY771306026 Key Organics / BIONET

(1R)-1-(2,6-dichlorophenyl)ethan-1-amine hydrochloride

  • MDL number MFCD12757835
  • Cas Number : 1131737-05-7
  • Molecular Weight : 226.53

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (1R)-1-(2,6-dichlorophenyl)ethanamine hydrochloride
Inchi Code 1S/C8H9Cl2N.ClH/c1-5(11)8-6(9)3-2-4-7(8)10;/h2-5H,11H2,1H3;1H/t5-;/m1./s1
Inchi Key QBWKCUFYMDZATP-NUBCRITNSA-N

Description

Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/665538

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/GS-7352_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart