KEY771306027 Key Organics / BIONET

(2S)-2-(2,2-difluoro-2H-1,3-benzodioxol-4-yl)pyrrolidine hydrochloride

  • MDL number MFCD26583537
  • Cas Number : 1212918-18-7
  • Molecular Weight : 263.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (S)-2-(2,2-difluorobenzo[d][1,3]dioxol-4-yl)pyrrolidine hydrochloride
Inchi Code 1S/C11H11F2NO2.ClH/c12-11(13)15-9-5-1-3-7(10(9)16-11)8-4-2-6-14-8;/h1,3,5,8,14H,2,4,6H2;1H/t8-;/m0./s1
Inchi Key BWVHGZWQWVNASS-QRPNPIFTSA-N

Description

Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/665539

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/GS-7353_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart