KEY771306032 Key Organics / BIONET

(1R)-1-(4-fluorophenyl)propan-1-amine hydrochloride

  • MDL number MFCD11101152
  • Cas Number : 1169576-95-7
  • Molecular Weight : 189.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (1R)-1-(4-fluorophenyl)-1-propanamine hydrochloride
Inchi Code 1S/C9H12FN.ClH/c1-2-9(11)7-3-5-8(10)6-4-7;/h3-6,9H,2,11H2,1H3;1H/t9-;/m1./s1
Inchi Key SNOAOHJQSMWUAH-SBSPUUFOSA-N

Description

Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/665544

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/GS-7363_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart