KEY771306034 Key Organics / BIONET

3-[(1S)-1-amino-2,2,2-trifluoroethyl]benzoic acid hydrochloride

  • MDL number MFCD12757419
  • Cas Number : 1391384-65-8
  • Molecular Weight : 255.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-[(1S)-1-amino-2,2,2-trifluoroethyl]benzoic acid hydrochloride
Inchi Code 1S/C9H8F3NO2.ClH/c10-9(11,12)7(13)5-2-1-3-6(4-5)8(14)15;/h1-4,7H,13H2,(H,14,15);1H/t7-;/m0./s1
Inchi Key PGVLYBVVIMPTQZ-FJXQXJEOSA-N

Description

Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/665546

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/GS-7365_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart