KEY771306036 Key Organics / BIONET

(2R)-2-[2-fluoro-5-(trifluoromethyl)phenyl]pyrrolidine hydrochloride

  • MDL number MFCD12758812
  • Cas Number : 1391408-54-0
  • Molecular Weight : 269.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/665548

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/GS-7369_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart