KEY771306037 Key Organics / BIONET

(2R)-2-[2-chloro-5-(trifluoromethyl)phenyl]pyrrolidine hydrochloride

  • MDL number MFCD12758788
  • Cas Number : 1391519-06-4
  • Molecular Weight : 286.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/665549

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/GS-7370_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart