KEY771306038 Key Organics / BIONET

2-[(2-oxo-2H-chromen-7-yl)oxy]acetic acid

  • MDL number MFCD00695616
  • Cas Number : 126424-85-9
  • Molecular Weight : 220.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name [(2-oxo-2H-chromen-7-yl)oxy]acetic acid
Inchi Code 1S/C11H8O5/c12-10(13)6-15-8-3-1-7-2-4-11(14)16-9(7)5-8/h1-5H,6H2,(H,12,13)
Inchi Key YENBPUAYKYLKSL-UHFFFAOYSA-N

Description

Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/664889

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/AS-3269_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart