KEY771306039 Key Organics / BIONET

(1S)-1-(pyridin-3-yl)ethan-1-amine dihydrochloride

  • MDL number MFCD16987845
  • Cas Number : 40154-84-5
  • Molecular Weight : 195.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (1S)-1-(3-pyridinyl)ethanamine dihydrochloride
Inchi Code 1S/C7H10N2.2ClH/c1-6(8)7-3-2-4-9-5-7;;/h2-6H,8H2,1H3;2*1H/t6-;;/m0../s1
Inchi Key FSDMOCFYBMVHLU-ILKKLZGPSA-N

Description

Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/665551

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/GS-7373_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart