KEY771306041 Key Organics / BIONET

2-chloro-N-(4-ethylphenyl)acetamide

  • MDL number MFCD00028177
  • Cas Number : 20172-36-5
  • Molecular Weight : 197.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-chloro-N-(4-ethylphenyl)acetamide
Inchi Code 1S/C10H12ClNO/c1-2-8-3-5-9(6-4-8)12-10(13)7-11/h3-6H,2,7H2,1H3,(H,12,13)
Inchi Key MNODWERQWKSALS-UHFFFAOYSA-N

Description

Purity 97%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/665552

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/JS-2985_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart