KEY771306047 Key Organics / BIONET

3-(dimethylamino)-2-methyl-1-phenylpropan-1-one hydrochloride

  • MDL number MFCD00034826
  • Cas Number : 5400-92-0
  • Molecular Weight : 227.73

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/665558

Safety Information

signalword Warning
pcodes P422, P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/JS-3001_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart