KEY771306049 Key Organics / BIONET

3-(dimethylamino)-1-(thiophen-2-yl)propan-1-one hydrochloride

  • MDL number MFCD00126349
  • Cas Number : 5424-47-5
  • Molecular Weight : 219.73

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/665560

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/JS-3003_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart