KEY771306060 Key Organics / BIONET

4-hydroxy-2-nitrobenzaldehyde

  • MDL number MFCD18397317
  • Cas Number : 90151-04-5
  • Molecular Weight : 167.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-hydroxy-2-nitrobenzaldehyde
Inchi Code 1S/C7H5NO4/c9-4-5-1-2-6(10)3-7(5)8(11)12/h1-4,10H
Inchi Key PTVFUHTYJLKEDZ-UHFFFAOYSA-N

Description

Purity 97%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/665571

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/JS-3016_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart