KEY771306072 Key Organics / BIONET

3-(dimethylamino)-1-(3-nitrophenyl)propan-1-one hydrochloride

  • MDL number MFCD01219754
  • Cas Number : 63352-94-3
  • Molecular Weight : 258.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/665583

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/JS-3030_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart