KEY771306075 Key Organics / BIONET

3-benzamidobicyclo[2.2.1]heptane-2-carboxylic acid

  • MDL number MFCD20085948
  • Cas Number : 1560390-24-0
  • Molecular Weight : 259.305

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/665586

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/JS-3033_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart