KEY771306078 Key Organics / BIONET

2-chloro-3-[(2,4-dimethoxyphenyl)amino]-1,4-dihydronaphthalene-1,4-dione

  • MDL number MFCD02986058
  • Cas Number : 378756-30-0
  • Molecular Weight : 343.77

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/665589

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/JS-3038_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart