KEY771306084 Key Organics / BIONET

2-(4-tert-butylphenyl)-4H-3,1-benzoxazin-4-one

  • MDL number MFCD00024127
  • Cas Number : 117145-55-8
  • Molecular Weight : 279.339

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/665595

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/JS-3048_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart