KEY771306087 Key Organics / BIONET

2-[(4,5-dibenzyl-4H-1,2,4-triazol-3-yl)sulfanyl]-N-[4-(morpholin-4-yl)phenyl]acetamide

  • MDL number MFCD30717702
  • Cas Number : 2085689-69-4
  • Molecular Weight : 499.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/665598

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/JS-3053_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart