KEY771306090 Key Organics / BIONET

N-(5-chloro-2-acetamido-4-nitrophenyl)acetamide

  • MDL number MFCD30717703
  • Cas Number : 154017-63-7
  • Molecular Weight : 271.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/665601

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/KM-0007_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart