KEY771306091 Key Organics / BIONET

N-(2-amino-4-chloro-5-nitrophenyl)-4-methylbenzene-1-sulfonamide

  • MDL number MFCD30717704
  • Cas Number : 2085689-70-7
  • Molecular Weight : 341.77

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/665602

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/KM-0008_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart