KEY771306093 Key Organics / BIONET

2,4-dichloro-5-(chloromethyl)pyridine

  • MDL number MFCD04114263
  • Cas Number : 73998-96-6
  • Molecular Weight : 196.46

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2,4-dichloro-5-(chloromethyl)pyridine
Inchi Code 1S/C6H4Cl3N/c7-2-4-3-10-6(9)1-5(4)8/h1,3H,2H2
Inchi Key PZHIYZLLIBBPFK-UHFFFAOYSA-N

Description

Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/665604

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/KM-0018_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart