KEY771306104 Key Organics / BIONET

(5Z)-5-{[2-(benzyloxy)-3-methoxyphenyl]methylidene}-2-sulfanylidene-1,3-thiazolidin-4-one

  • MDL number MFCD02377000
  • Cas Number : 1287651-60-8
  • Molecular Weight : 357.44

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 182-184 deg C (Lit.)
Purity 97
Storage temp. Store at 4
Shipping Temperature Store at 4
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/665612

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404,P411
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/NS-00055_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart