KEY771306105 Key Organics / BIONET

methyl 3-chloro-5-methoxy-1-benzothiophene-2-carboxylate

  • MDL number MFCD00100498
  • Cas Number : 41280-81-3
  • Molecular Weight : 256.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 113-115 deg C (Lit.)
Purity 97
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/665613

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P302 + P352,P304 + P340,P305 + P351 + P338,P402 + P404
hcodes H302,H312,H315,H319,H332,H335
MSDS https://www.keyorganics.net/media/catalog/product/sds/NS-00056_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart