KEY771306108 Key Organics / BIONET

3-[(4-methoxyphenyl)amino]-1-phenylpropan-1-one

  • MDL number MFCD00156684
  • Cas Number : 4173-65-3
  • Molecular Weight : 255.317

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 110-112 deg C (lit.)
Purity 95
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/665616

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/NS-00059_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart