KEY771306118 Key Organics / BIONET

3-(piperidin-2-yl)-1H-indole

  • MDL number MFCD02663593
  • Cas Number : 4695-73-2
  • Molecular Weight : 200.285

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/665626

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/PS-0004_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart