KEY771306120 Key Organics / BIONET

tert-butyl 2-[(4-methoxyphenyl)methyl]-3,5-dioxo-2,9-diazaspiro[5.5]undecane-9-carboxylate

  • MDL number MFCD30490120
  • Cas Number : 1312814-90-6
  • Molecular Weight : 402.49

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/440298

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/SS-4852_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart