KEY771306122 Key Organics / BIONET

3-[(benzyloxy)methyl]pentane-1,5-diol

  • MDL number MFCD30717729
  • Cas Number : 1935241-67-0
  • Molecular Weight : 224.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/665628

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/SS-4944_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart