KEY771306130 Key Organics / BIONET

1,4-bis(thiophen-2-yl)butane-1,4-dione

  • MDL number MFCD00208362
  • Cas Number : 13669-05-1
  • Molecular Weight : 250.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/665636

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/SS-4971_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart