KEY771306135 Key Organics / BIONET

(R)-1-(2-carboxynaphthalen-1-yl)naphthalene-2-carboxylic acid

  • MDL number MFCD00185729
  • Cas Number : 80703-23-7
  • Molecular Weight : 342.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/665641

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/SS-4976_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart