KEY947334813 Key Organics / BIONET

ethyl 1-(benzenesulfonyl)piperidine-4-carboxylate

  • MDL number MFCD00591722
  • Cas Number : 111627-26-0
  • Molecular Weight : 297.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/65704

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/1R-1328_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart