KEY947334814 Key Organics / BIONET

2-(trifluoromethyl)-1H-1,3-benzodiazole

  • MDL number MFCD00022675
  • Cas Number : 312-73-2
  • Molecular Weight : 186.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 207 - 212
Iupac Name 2-(trifluoromethyl)-1H-benzimidazole
Inchi Code 1S/C8H5F3N2/c9-8(10,11)7-12-5-3-1-2-4-6(5)13-7/h1-4H,(H,12,13)
Inchi Key MXFMPTXDHSDMTI-UHFFFAOYSA-N

Description

Melting Point 207 - 212
Purity 98%
Storage temp. Store at 4
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/65718

Safety Information

signalword Danger
pcodes P260,P262,P270,P280,P302 + P352,P304 + P340,P305 + P351 + P338,P402 + P404,P411
MSDS https://www.keyorganics.net/media/catalog/product/sds/1R-1542_SDS.pdf
hcodes H300,H311,H319,H331,H335
pictograms GHS06
Availability details for
Quantity
  • Availability
  • Add to Cart