KEY947334854 Key Organics / BIONET

4-chloro-2-(methylsulfanyl)thieno[3,2-d]pyrimidine

  • MDL number MFCD01444000
  • Cas Number : 176530-47-5
  • Molecular Weight : 216.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-chloro-2-(methylsulfanyl)thieno[3,2-d]pyrimidine
Inchi Code 1S/C7H5ClN2S2/c1-11-7-9-4-2-3-12-5(4)6(8)10-7/h2-3H,1H3
Inchi Key YKVAWSVTEWXJGJ-UHFFFAOYSA-N

Description

Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/83147

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/6N-420S_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart