KEY947334855 Key Organics / BIONET

2-methyl-[1,3]oxazolo[4,5-b]pyridine

  • MDL number MFCD05256196
  • Cas Number : 86467-39-2
  • Molecular Weight : 134.138

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/84365

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/6X-0823_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart