KEY947334860 Key Organics / BIONET

4-chloro-2-(methylsulfanyl)-5,6,7,8-tetrahydroquinazoline

  • MDL number MFCD01568403
  • Cas Number : 51660-11-8
  • Molecular Weight : 214.72

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-chloro-2-(methylsulfanyl)-5,6,7,8-tetrahydroquinazoline
Inchi Code 1S/C9H11ClN2S/c1-13-9-11-7-5-3-2-4-6(7)8(10)12-9/h2-5H2,1H3
Inchi Key XDQBZDZLOKZGBN-UHFFFAOYSA-N

Description

Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/86788

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/7N-430S_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart